5-(tert-Butoxycarbonyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A136883-1G A136883-250MG
77010-166EA 165.51 USD
77010-166 77010-168
5-(tert-Butoxycarbonyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine-2-carboxylic acid 95%
5-(tert-Butoxycarbonyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₄S
MW: 283.34 g/mol
MDL Number: MFCD08448165
CAS Number: 165947-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR