1-Fluoro-3-pentylbenzene 98%

Supplier: Ambeed
Warning

A136723-1G A136723-5G
76984-020EA 46.68 USD
76984-020 76984-022
1-Fluoro-3-pentylbenzene 98%
1-Fluoro-3-pentylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅F
MW: 166.23 g/mol
MDL Number: MFCD22376645
CAS Number: 28593-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR