4-(4-Methanesulfonylpiperazin-1-yl)phenylamine 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-(Methylsulphonyl)piperazin-1-yl)aniline, 4-[4-(Methylsulphonyl)piperazin-1-yl]aniline

A136426-100MG A136426-250MG
76996-672EA 14.86 USD
76996-672 76996-674
4-(4-Methanesulfonylpiperazin-1-yl)phenylamine 98%
4-(4-Methanesulfonylpiperazin-1-yl)phenylamine

Precaución: for Research Use Only

Formula: C₁₁H₁₇N₃O₂S
MW: 255.34 g/mol
MDL Number: MFCD03848236
CAS Number: 442549-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR