3-Amino-2-bromo-6-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-methoxypyridin-3-amine

A136398-10G A136398-1G A136398-250MG A136398-25G A136398-5G
76996-662EA 80 USD
76996-662 76996-664 76996-666 76996-668 76996-670
3-Amino-2-bromo-6-methoxypyridine 98%
3-Amino-2-bromo-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MDL Number: MFCD05662394
CAS Number: 135795-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR