(2R,3R)-3-((S)-1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-dolaproine

A136386-100MG A136386-250MG A136386-1G
76984-014EA 66.84 USD
76984-014 76984-016 77683-204
(2R,3R)-3-((S)-1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoic acid 95%
(2R,3R)-3-((S)-1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoic acid

Precaución: For research use only.

Formula: C₁₄H₂₅NO₅
MW: 287.352 g/mol
MDL Number: MFCD18157735
CAS Number: 120205-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR