Methyl-3-amino-5-fluorobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-amino-5-fluorobenzoate
A136109-100MG
A136109-10G
A136109-1G
A136109-250MG
A136109-5G
77027-316EA
6
USD
77027-316
77027-318
77027-320
77027-322
77027-324
Methyl-3-amino-5-fluorobenzoate 98%
Methyl-3-amino-5-fluorobenzoate
Precaución: for Research Use Only
Formula:
C₈H₈FNO₂ MW: 169.16 g/mol |
MDL Number:
MFCD05863683 CAS Number: 884497-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...