2-[3-(Bromomethyl)-5-(1-cyano-1-methylethyl)phenyl]-2-methylpropanenitrile 95%

Supplier: Ambeed
Warning

Synonyms: 2,2'-(5-(Bromomethyl)-1,3-phenylene)bis(2-methylpropanenitrile)

A136077-10G A136077-1G A136077-250MG A136077-25G A136077-5G
76996-638EA 163.38 USD
76996-638 76996-640 76996-642 76996-644 76996-646
2-[3-(Bromomethyl)-5-(1-cyano-1-methylethyl)phenyl]-2-methylpropanenitrile 95%
2-[3-(Bromomethyl)-5-(1-cyano-1-methylethyl)phenyl]-2-methylpropanenitrile

Precaución: for Research Use Only

Formula: C₁₅H₁₇BrN₂
MW: 305.23 g/mol
MDL Number: MFCD07782114
CAS Number: 120511-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR