1-Benzyl-4-methylpiperidin-3-ol 95%
Supplier: Ambeed
|
Warning
|
A136069-250MG
A136069-5G
77027-312EA
50.93
USD
77027-312
77027-314
1-Benzyl-4-methylpiperidin-3-ol 95%
1-Benzyl-4-methylpiperidin-3-ol
Precaución: for Research Use Only
Formula:
C₁₃H₁₉NO MW: 205.3 g/mol |
MDL Number:
MFCD16619305 CAS Number: 384338-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...