3-Bromo-1,5-dimethyl-1H-pyrazole 97%

Supplier: Ambeed
Warning

A136043-1G A136043-250MG A136043-5G
77010-142EA 71.09 USD
77010-142 77010-144 77010-146
3-Bromo-1,5-dimethyl-1H-pyrazole 97%
3-Bromo-1,5-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₇BrN₂
MW: 175.026 g/mol
MDL Number: MFCD09991542
CAS Number: 5744-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR