Benzyl 2,6-dimethyl-4-oxopiperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A135817-100MG A135817-1G A135817-250MG
76983-980EA 221 USD
76983-980 76983-982 76983-984
Benzyl 2,6-dimethyl-4-oxopiperidine-1-carboxylate 97%
Benzyl 2,6-dimethyl-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₃
MDL Number: MFCD16250702
CAS Number: 1233932-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR