tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
tert-Butyl 3-(hydroxymethyl)pyrrolidine-1-carboxylate, 1-BOC-3-Pyrrolidinemethanol, 1-Boc-3-(hydroxymethyl)pyrrolidine
A135811-100G
A135811-10G
A135811-250MG
A135811-25G
A135811-5G
A135811-1G
77010-130EA
490.14
USD
77010-130
77010-132
77010-134
77010-136
77010-138
77592-560
tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate 97%
tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₃ MW: 201.27 g/mol |
MDL Number:
MFCD02179040 CAS Number: 114214-69-6 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...