4-Bromo-N1-methylbenzene-1,2-diamine 97%

Supplier: Ambeed
Warning

A135762-10G A135762-1G A135762-250MG A135762-5G
77010-122EA 413.76 USD
77010-122 77010-124 77010-126 77010-128
4-Bromo-N1-methylbenzene-1,2-diamine 97%
4-Bromo-N1-methylbenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MW: 201.06 g/mol
MDL Number: MFCD10693151
CAS Number: 69038-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR