5-Fluoro-4-iodo-2-methylaniline 95+%

Supplier: Ambeed
Warning

A135701-250MG A135701-5G
76983-974EA 20 USD
76983-974 76983-976
5-Fluoro-4-iodo-2-methylaniline 95+%
5-Fluoro-4-iodo-2-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇FIN
MDL Number: MFCD21364952
CAS Number: 307306-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR