2-Bromo-N-[3-(trifluoromethyl)phenyl]acetamide 98%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-N-(3-(trifluoromethyl)phenyl)acetamide, 2-Bromo-3'-(trifluoromethyl)acetanilide

A135677-100MG A135677-1G A135677-250MG A135677-5G
76996-622EA 5 USD
76996-622 76996-624 76996-626 76996-628
2-Bromo-N-[3-(trifluoromethyl)phenyl]acetamide 98%
2-Bromo-N-[3-(trifluoromethyl)phenyl]acetamide

Precaución: for Research Use Only

Formula: C₉H₇BrF₃NO
MDL Number: MFCD00859566
CAS Number: 25625-57-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR