3-Bromo-5-fluoro-1H-indazole 95%

Supplier: Ambeed
Warning

A135574-100MG A135574-250MG
77027-294EA 27.59 USD
77027-294 77027-296
3-Bromo-5-fluoro-1H-indazole 95%
3-Bromo-5-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MW: 215.0200 g/mol
MDL Number: MFCD07781639
CAS Number: 885519-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR