2,3,5-Tribromo-4-methylthiophene 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-2,4,5-tribromothiophene

A135570-1G A135570-250MG A135570-25G A135570-5G
76983-966EA 6.37 USD
76983-966 76983-968 76983-970 76983-972
2,3,5-Tribromo-4-methylthiophene 95%
2,3,5-Tribromo-4-methylthiophene

Precaución: for Research Use Only

Formula: C₅H₃Br₃S
MW: 334.85 g/mol
MDL Number: MFCD00130086
CAS Number: 67869-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR