Fmoc-Dopa(acetonide)-OH 97%

Supplier: Ambeed
Warning

A135438-1G A135438-250MG A135438-5G A135438-100MG
77010-110EA 266.29 USD
77010-110 77010-112 77566-656 77683-199
Fmoc-Dopa(acetonide)-OH 97%
Fmoc-Dopa(acetonide)-OH

Precaución: For research use only.

Formula: C₂₇H₂₅NO₆
MW: 459.5 g/mol
MDL Number: MFCD08064332
CAS Number: 852288-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR