5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulfonamide 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-((4-((2,3-Dimethyl-2H-indazol-6-yl)(methyl)amino)pyrimidin-2-yl)amino)-2-methylbenzenesulphonamide, Pazopanib
A135375-50MG
A135375-100MG
A135375-1G
A135375-250MG
A135375-1MG
77027-280EA
8.49
USD
77027-280
77599-962
77599-964
77599-966
77666-956
5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulfonamide 99%
5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulfonamide
Precaución: for Research Use Only
Formula:
C₂₁H₂₃N₇O₂S MW: 437.53 g/mol |
MDL Number:
MFCD11616589 CAS Number: 444731-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...