(±)-2-Pyrrolidinone-5-carboxylic acid 99%

Supplier: Ambeed
Warning

A135319-1000G A135319-100G A135319-500G A135319-25G
76996-606EA 48 USD
76996-606 76996-608 76996-610 77314-942
(±)-2-Pyrrolidinone-5-carboxylic acid 99%
(±)-2-Pyrrolidinone-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₇NO₃
MDL Number: MFCD00064322
CAS Number: 149-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR