1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-1,3,5-triazinane-2,4,6-trione 95%

Supplier: Ambeed
Warning

Synonyms: Isocyanuric acid tris(3,5-di-tert-butyl-4-hydroxybenzyl) ester, Tris(3,5-di-tert-butyl-4-hydroxybenzyl) Isocyanurate

A135297-1000G A135297-25G A135297-500G A135297-100G
76996-600EA 55.17 USD
76996-600 76996-602 76996-604 77670-944
1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-1,3,5-triazinane-2,4,6-trione 95%
1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-1,3,5-triazinane-2,4,6-trione

Precaución: for Research Use Only

Formula: C₄₈H₆₉N₃O₆
MW: 784.10 g/mol
MDL Number: MFCD00134700
CAS Number: 27676-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR