Methyl-5-bromo-1H-indazole-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromo-1H-indazole-3-carboxylate

A135219-100MG A135219-10G A135219-1G A135219-250MG A135219-25G A135219-5G
77010-098EA 7 USD
77010-098 77010-100 77010-102 77010-104 77010-106 77010-108
Methyl-5-bromo-1H-indazole-3-carboxylate 95%
Methyl 5-bromo-1H-indazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MDL Number: MFCD07371572
CAS Number: 78155-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR