Methyl 6-chloro-1H-indole-4-carboxylate 97%

Supplier: Ambeed
Warning

A135103-100MG A135103-1G A135103-250MG
76983-948EA 13.8 USD
76983-948 76983-950 76983-952
Methyl 6-chloro-1H-indole-4-carboxylate 97%
Methyl 6-chloro-1H-indole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO₂
MW: 209.62 g/mol
MDL Number: MFCD11845461
CAS Number: 1082040-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR