Methyl 2,5-dibromothiophene-3-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A135077-100MG
A135077-1G
A135077-250MG
77010-092EA
13.8
USD
77010-092
77010-094
77010-096
Methyl 2,5-dibromothiophene-3-carboxylate 95%
Methyl 2,5-dibromothiophene-3-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₄Br₂O₂S MW: 299.97 g/mol |
CAS Number:
89280-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...