2-Amino-6-chloroimidazo[1,2-b]pyridazine 97%

Supplier: Ambeed
Warning

A135022-100MG A135022-250MG A135022-25G A135022-5G A135022-1G
76996-588EA 11 USD
76996-588 76996-590 76996-592 76996-594 77670-940
2-Amino-6-chloroimidazo[1,2-b]pyridazine 97%
2-Amino-6-chloroimidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C₆H₅ClN₄
MDL Number: MFCD16495902
CAS Number: 887625-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR