Methyl 2-phenylpropanoate 98%

Supplier: Ambeed
Warning

A134981-10G A134981-1G
77010-088EA 112.46 USD
77010-088 77010-090
Methyl 2-phenylpropanoate 98%
Methyl 2-phenylpropanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
MDL Number: MFCD06795946
CAS Number: 31508-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR