L-Serine ethyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Ser-OEt.HCl, H-Ser-OEt·HCl

A134900-100G A134900-1G A134900-25G A134900-500G A134900-5G A134900-10G
76983-930EA 39.26 USD
76983-930 76983-932 76983-934 76983-936 76983-938 77611-210
L-Serine ethyl ester hydrochloride 97%
L-Serine ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO₃
MW: 169.605 g/mol
MDL Number: MFCD00012594
CAS Number: 26348-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR