3-Benzyl-1-dodecyl-2-methyl-1H-imidazol-3-ium chloride 97%

Supplier: Ambeed
Warning

A134756-1G A134756-250MG A134756-5G
77027-250EA 8.49 USD
77027-250 77027-252 77027-254
3-Benzyl-1-dodecyl-2-methyl-1H-imidazol-3-ium chloride 97%
3-Benzyl-1-dodecyl-2-methyl-1H-imidazol-3-ium chloride

Precaución: for Research Use Only

Formula: C₂₃H₃₇ClN₂
MW: 377.00 g/mol
MDL Number: MFCD00050314
CAS Number: 21054-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR