2-Bromo-5-chloro-4-methylaniline 95%

Supplier: Ambeed
Warning

A134733-100MG A134733-250MG
77010-082EA 11.67 USD
77010-082 77010-084
2-Bromo-5-chloro-4-methylaniline 95%
2-Bromo-5-chloro-4-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrClN
MW: 220.4942 g/mol
MDL Number: MFCD00806533
CAS Number: 102170-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR