2,5-Dibromophenylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2,5-Dibromophenyl)acetic acid

A134728-100G A134728-10G A134728-1G A134728-250MG A134728-25G A134728-5G
77027-240EA 301.3 USD
77027-240 77027-242 77027-244 77027-246 77027-248 77670-932
2,5-Dibromophenylacetic acid 98%
2,5-Dibromophenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
MW: 293.95 g/mol
MDL Number: MFCD02664691
CAS Number: 203314-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR