5-Bromo-3-[(trimethylsilyl)ethynyl]pyrazin-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-((trimethylsilyl)ethynyl)pyrazin-2-amine

A134711-1G A134711-250MG
77027-236EA 68.96 USD
77027-236 77027-238
5-Bromo-3-[(trimethylsilyl)ethynyl]pyrazin-2-amine 97%
5-Bromo-3-[(trimethylsilyl)ethynyl]pyrazin-2-amine

Precaución: for Research Use Only

Formula: C₉H₁₂BrN₃Si
MW: 270.201 g/mol
MDL Number: MFCD16660032
CAS Number: 875781-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR