3-Bromoimidazo[1,2-b]pyridazin-6-ylamine 95%

Supplier: Ambeed
Warning

A134690-100MG A134690-250MG
77027-228EA 49 USD
77027-228 77027-232
3-Bromoimidazo[1,2-b]pyridazin-6-ylamine 95%
3-Bromoimidazo[1,2-b]pyridazin-6-ylamine

Precaución: for Research Use Only

Formula: C6H5BrN4
MDL Number: MFCD11520885
CAS Number: 1260850-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR