1-(Cyclopropylcarbonyl)-4-[5-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoyl]piperazine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Olaparib, 4-(3-(4-(Cyclopropanecarbonyl)piperazine-1-carbonyl)-4-fluorobenzyl)phthalazin-1(2H)-one
A134565-10MG
A134565-1G
A134565-5G
A134565-100MG
A134565-250MG
77027-216EA
5.31
USD
77027-216
77027-218
77027-220
77599-942
77599-944
1-(Cyclopropylcarbonyl)-4-[5-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoyl]piperazine 98%
1-(Cyclopropylcarbonyl)-4-[5-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoyl]piperazine
Precaución: for Research Use Only
Formula:
C₂₄H₂₃FN₄O₃ MW: 434.47 g/mol |
MDL Number:
MFCD13185161 CAS Number: 763113-22-0 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...