(5-Methoxybenzofuran-2-yl)boronic acid 95%

Supplier: Ambeed
Warning

A134230-5G A134230-1G A134230-250MG A134230-100MG
76983-894EA 774 USD
76983-894 77566-630 77611-202 77889-911
(5-Methoxybenzofuran-2-yl)boronic acid 95%
(5-Methoxybenzofuran-2-yl)boronic acid

Precaución: For research use only.

Formula: C₉H₉BO₄
MDL Number: MFCD06801701
CAS Number: 551001-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR