tert-Butyl-7-azaspiro[3.5]nonan-2-ylcarbamate 97%

Supplier: Ambeed
Warning

A134198-100MG A134198-1G A134198-250MG A134198-5G
77027-210EA 26 USD
77027-210 77027-212 77027-214 77666-940
tert-Butyl-7-azaspiro[3.5]nonan-2-ylcarbamate 97%
tert-Butyl-7-azaspiro[3.5]nonan-2-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₂
MDL Number: MFCD12198553
CAS Number: 147611-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR