N,N',N''-Tris(p-toluenesulfonyl)diethylenetriamine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-N,N-bis(2-(4-methylphenylsulphonamido)ethyl)benzenesulphonamide, N,N',N''-Tritosyldiethylenetriamine

A134190-100G A134190-10G A134190-1G A134190-250MG A134190-5G A134190-25G
76996-554EA 47 USD
76996-554 76996-556 76996-558 76996-560 76996-562 77881-064
N,N',N''-Tris(p-toluenesulfonyl)diethylenetriamine 95%
N,N',N''-Tris(p-toluenesulfonyl)diethylenetriamine

Precaución: for Research Use Only

Formula: C₂₅H₃₁N₃O₆S₃
MDL Number: MFCD00015623
CAS Number: 56187-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR