Ethyl (4-fluorobenzoyl)acetate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-(4-Fluorophenyl)-3-oxopropionate, 3-(4-Fluorophenyl)-3-oxopropionic Acid Ethyl Ester, (4-Fluorobenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(4-fluorophenyl)-3-oxopropanoate

A134139-100G A134139-10G A134139-1G A134139-25G A134139-5G
77010-064EA 195 USD
77010-064 77010-066 77010-068 77611-198 77611-200
Ethyl (4-fluorobenzoyl)acetate 96%
Ethyl (4-fluorobenzoyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₃
MDL Number: MFCD03093631
CAS Number: 1999-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR