Boc-L-allylglycine . DCHA 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Allyl-Gly-OH·DCHA, Dicyclohexylamine (S)-2-((tert-butoxycarbonyl)amino)pent-4-enoate, (S)-N-BOC-alpha-Allylglycine dicyclohexylamine salt

A134114-1G A134114-25G A134114-5G
76996-552EA 10.61 USD
76996-552 77670-918 77670-920
Boc-L-allylglycine . DCHA 97%
Boc-L-allylglycine . DCHA

Precaución: for Research Use Only

Formula: C₂₂H₄₀N₂O₄
MW: 396.58 g/mol
MDL Number: MFCD01321013
CAS Number: 143979-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR