4-(2-Pyridyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Pyridin-2-yl)phenylboronic acid, (4-(Pyridin-2-yl)phenyl)boronic acid

A134063-10G A134063-1G A134063-250MG A134063-5G
76983-886EA 561.22 USD
76983-886 76983-888 76983-890 76983-892
4-(2-Pyridyl)phenylboronic acid 98%
4-(2-Pyridyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀BNO₂
MW: 199.014 g/mol
MDL Number: MFCD17013972
CAS Number: 170230-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR