9,9-Diphenylfluorene-2-boronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(9,9-Diphenyl-9H-fluoren-2-yl)boronic acid
A133858-10G
A133858-1G
A133858-5G
77010-042EA
240.83
USD
77010-042
77010-044
77010-046
9,9-Diphenylfluorene-2-boronic acid 98%
9,9-Diphenylfluorene-2-boronic acid
Precaución: for Research Use Only
Formula:
C₂₅H₁₉BO₂ MW: 362.24 g/mol |
MDL Number:
MFCD22581299 CAS Number: 400607-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...