3-Bromo-2-isopropoxypyridine 95%

Supplier: Ambeed
Warning

A133823-1G A133823-5G
76983-862EA 17 USD
76983-862 77889-905
3-Bromo-2-isopropoxypyridine 95%
3-Bromo-2-isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MDL Number: MFCD09835265
CAS Number: 717843-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR