Valsartan 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(N-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid, (S)-2-(N-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid

A133780-50MG A133780-10G A133780-100G A133780-25G
76996-544EA 5.31 USD
76996-544 77549-606 77599-922 77599-924
Valsartan 95%
Valsartan

Precaución: for Research Use Only

Formula: C₂₄H₂₉N₅O₃
MW: 435.52 g/mol
MDL Number: MFCD00865840
CAS Number: 137862-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR