3-Fluoro-2-methylphenylacetonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 2-(3-Fluoro-2-methylphenyl)acetonitrile

A133598-1G A133598-250MG A133598-25G
76983-848EA 50.93 USD
76983-848 76983-850 76983-852
3-Fluoro-2-methylphenylacetonitrile 95%
3-Fluoro-2-methylphenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₈FN
MW: 149.17 g/mol
MDL Number: MFCD03788544
CAS Number: 500912-15-2
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR