1-(2-Bromo-6-methoxyphenyl)ethanone 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-methoxyacetophenone

A133523-1G A133523-250MG A133523-25G A133523-5G
77010-034EA 33.95 USD
77010-034 77010-036 77010-038 77010-040
1-(2-Bromo-6-methoxyphenyl)ethanone 97%
1-(2-Bromo-6-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD22573041
CAS Number: 380225-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR