5-(3-Fluorophenyl)-2-methylpyridine 97%

Supplier: Ambeed
Warning

A133505-10G A133505-1G A133505-250MG A133505-25G
77027-184EA 89 USD
77027-184 77027-186 77027-188 77027-190
5-(3-Fluorophenyl)-2-methylpyridine 97%
5-(3-Fluorophenyl)-2-methylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀FN
MW: 187.213 g/mol
MDL Number: MFCD13152238
CAS Number: 713143-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR