(S)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine 98%

Supplier: Ambeed
Warning

A133440-1G A133440-250MG A133440-5G A133440-25G
76983-842EA 20.16 USD
76983-842 76983-844 76983-846 77611-188
(S)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine 98%
(S)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₄S
MW: 273.349 g/mol
MDL Number: MFCD22677268
CAS Number: 608141-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR