6-(Methylamino)-3-pyridinyl boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

A133254-100MG A133254-1G A133254-250MG A133254-5G
77027-166EA 55.17 USD
77027-166 77027-168 77027-170 77027-172
6-(Methylamino)-3-pyridinyl boronic acid pinacol ester 98%
6-(Methylamino)-3-pyridinyl boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₉BN₂O₂
MW: 234.102 g/mol
MDL Number: MFCD11878220
CAS Number: 1005009-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR