7H-Pyrrolo[2,3-c]pyridazine 95%

Supplier: Ambeed
Warning

A133172-250MG A133172-100MG
77027-156EA 162 USD
77027-156 77871-626
7H-Pyrrolo[2,3-c]pyridazine 95%
7H-Pyrrolo[2,3-c]pyridazine

Precaución: for Research Use Only

Formula: C₆H₅N₃
MDL Number: MFCD11110436
CAS Number: 16767-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR