Ethyl-4-amino-2-chlorobenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-amino-2-chlorobenzoate

A132883-250MG A132883-5G
76996-484EA 29.71 USD
76996-484 76996-486
Ethyl-4-amino-2-chlorobenzoate 95%
Ethyl 4-amino-2-chlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.6342 g/mol
MDL Number: MFCD09034640
CAS Number: 16017-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR