Ethyl-4-amino-2-chloropyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

A132737-10G A132737-250MG A132737-5G A132737-1G
77027-140EA 250 USD
77027-140 77027-142 77027-144 77666-906
Ethyl-4-amino-2-chloropyrimidine-5-carboxylate 97%
Ethyl-4-amino-2-chloropyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈ClN₃O₂
MDL Number: MFCD09860847
CAS Number: 71406-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR