(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
PyBOP, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate
A132694-10G
A132694-5G
A132694-25G
A132694-100G
77027-132EA
8.49
USD
77027-132
77027-134
77314-344
77549-574
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 98%
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Precaución: for Research Use Only
Formula:
C₁₈H₂₈F₆N₆OP₂ MW: 520.4 g/mol |
MDL Number:
MFCD00077411 CAS Number: 128625-52-5 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...